Learn More
3-(Dimethylsulfamoyl)benzeneboronic acid, 98%
CAS: 871329-59-8 | C8H12BNO4S | 229.057 g/mol
Supplier: Thermo Scientific Chemicals H5283803
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-(Dimethylsulfamoyl)benzeneboronic acid | |
| 88°C to 92°C | |
| MFCD07363747 | |
| 3-n,n-dimethylsulfamoyl phenyl boronic acid, n,n-dimethyl 3-boronobenzenesulfonamide, 3-dimethylsulfamoyl phenyl boronic acid, 3-n,n-dimethylsulphonamido benzeneboronic acid, 3-dimethylsulfamoyl phenylboronic acid, 3-dimethylsulfamoyl benzeneboronic acid, 3-n,n-dimethylsulfonamidophenyl boronic acid, boronic acid, 3-dimethylamino sulfonyl phenyl, boronic acid,b-3-dimethylamino sulfonyl phenyl, acmc-209qg4 | |
| B(C1=CC(=CC=C1)S(=O)(=O)N(C)C)(O)O | |
| 229.057 | |
| 229.06 |
| 871329-59-8 | |
| C8H12BNO4S | |
| 1 g | |
| SGERXKCDWJPIOS-UHFFFAOYSA-N | |
| [3-(dimethylsulfamoyl)phenyl]boronic acid | |
| 44119634 | |
| 98% |
Chemical Identifiers
| 871329-59-8 | |
| 229.057 | |
| SGERXKCDWJPIOS-UHFFFAOYSA-N | |
| 44119634 | |
| B(C1=CC(=CC=C1)S(=O)(=O)N(C)C)(O)O |
| C8H12BNO4S | |
| MFCD07363747 | |
| 3-n,n-dimethylsulfamoyl phenyl boronic acid, n,n-dimethyl 3-boronobenzenesulfonamide, 3-dimethylsulfamoyl phenyl boronic acid, 3-n,n-dimethylsulphonamido benzeneboronic acid, 3-dimethylsulfamoyl phenylboronic acid, 3-dimethylsulfamoyl benzeneboronic acid, 3-n,n-dimethylsulfonamidophenyl boronic acid, boronic acid, 3-dimethylamino sulfonyl phenyl, boronic acid,b-3-dimethylamino sulfonyl phenyl, acmc-209qg4 | |
| [3-(dimethylsulfamoyl)phenyl]boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only