missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)-1-phenyl-2-propen-1-one 98.0+%, TCI America™

Catalog No. D53841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53841G 1 g
D53845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53841G Supplier TCI America Supplier No. D53841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1201-93-0
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD03939120
InChI Key HUTKDPINCSJXAA-CMDGGOBGSA-N
Synonym 3-(Dimethylamino)acrylophenone
PubChem CID 5398495
IUPAC Name (E)-3-(dimethylamino)-1-phenylprop-2-en-1-one
SMILES CN(C)C=CC(=O)C1=CC=CC=C1

Specifications

Melting Point 92°C
Quantity 1 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)-1-phenyl-2-propen-1-one