accessibility menu, dialog, popup

UserName

CH3NH3CO(E/Z)

CAS RN 1201-93-0

IUPAC Name: 3-(dimethylamino)-1-phenylprop-2-en-1-one
Synonyms: 3-(dimethylamino)-1-phenylprop-2-en-1-one
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: HUTKDPINCSJXAA-UHFFFAOYSA-N
SMILES: CN(C)C=CC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results