missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 3'-Deoxythymidine
CAS: 3416-05-5 | C10H14N2O4 | 226.23 g/mol
$779.95 - $1,373.62
Chemical Identifiers
| CAS | 3416-05-5 |
|---|---|
| Molecular Formula | C10H14N2O4 |
| Molecular Weight (g/mol) | 226.23 |
| MDL Number | MFCD00010570 |
| InChI Key | XKKCQTLDIPIRQD-JGVFFNPUSA-N |
| Synonym | 3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf |
| PubChem CID | 65119 |
| IUPAC Name | 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
| SMILES | CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ65765ME
|
Thermo Scientific Chemicals
J65765ME |
500 mg |
Each for $779.95
|
|
|||||
|
AAJ6576503
|
Thermo Scientific Chemicals
J6576503 |
1 g |
Each for $1,373.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C10H14N2O4 | |
| MFCD00010570 | |
| 3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf | |
| 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
Specifications
| 3416-05-5 | |
| White to Yellow | |
| MFCD00010570 | |
| 3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf | |
| CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O | |
| 226.23 | |
| 226.23 | |
| 3'-Deoxythymidine |
| 155°C to 156°C | |
| C10H14N2O4 | |
| 21884 | |
| XKKCQTLDIPIRQD-JGVFFNPUSA-N | |
| 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione | |
| 65119 | |
| Powder |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only