missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Deoxythymidine

Catalog No. AAJ6576503
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6576503 1 g
AAJ65765ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6576503 Supplier Thermo Scientific Chemicals Supplier No. J6576503
Only null left

CAS: 3416-05-5 | C10H14N2O4 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3416-05-5
Molecular Formula C10H14N2O4
Molecular Weight (g/mol) 226.23
MDL Number MFCD00010570
InChI Key XKKCQTLDIPIRQD-JGVFFNPUSA-N
Synonym 3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf
PubChem CID 65119
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O

Specifications

Melting Point 155°C to 156°C
Color White to Yellow
Quantity 1 g
Beilstein 21884
Formula Weight 226.23
Physical Form Powder
Chemical Name or Material 3'-Deoxythymidine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only