missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Deoxythymidine

CAS: 3416-05-5 | C10H14N2O4 | 226.23 g/mol

Supplier:  Thermo Scientific Chemicals J65765ME

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ65765ME


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

3'-Deoxythymidine
155°C to 156°C
C10H14N2O4
500 mg
3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf
CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O
226.23
226.23
3416-05-5
White to Yellow
MFCD00010570
21884
XKKCQTLDIPIRQD-JGVFFNPUSA-N
1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
65119
Powder

Identifiants chimiques

3416-05-5
226.23
XKKCQTLDIPIRQD-JGVFFNPUSA-N
65119
CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O
C10H14N2O4
MFCD00010570
3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf
1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only