missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine, 98%

CAS: 100-54-9 | C6H4N2 | 104.11 g/mol

Supplier:  Thermo Scientific Chemicals A148500E

 View more versions of this product

Catalog No. AAA148500E


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Cyanopyridine
48°C to 51°C
206°C to 208°C
Pungent
MFCD00006372
107711
GZPHSAQLYPIAIN-UHFFFAOYSA-N
pyridine-3-carbonitrile
79
104.11
100-54-9
1.16
84°C (183°F)
C6H4N2
2500 g
3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
N#CC1=CC=CN=C1
104.11
CHEBI:86556
98%

Chemical Identifiers

100-54-9
104.11
GZPHSAQLYPIAIN-UHFFFAOYSA-N
79
pyridine-3-carbonitrile
C6H4N2
MFCD00006372
3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
CHEBI:86556
N#CC1=CC=CN=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-863-0

RTECSNumber : QT3030000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only