accessibility menu, dialog, popup

UserName

NN

CAS RN 100-54-9

IUPAC Name: pyridine-3-carbonitrile
Synonyms: 3-cyanopyridine
Molecular Weight (g/mol): 104.11
Molecular Formula: C6H4N2
InChi Key: GZPHSAQLYPIAIN-UHFFFAOYSA-N
SMILES: N#CC1=CC=CN=C1
Boiling Point 
  • (2)
  • (1)
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (7)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (7)
  • (4)
  • (2)
  • (1)