missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanopropionaldehyde Dimethyl Acetal 98.0+%, TCI America™
$238.60 - $1427.87
Chemical Identifiers
| CAS | 14618-78-1 |
|---|---|
| Molecular Formula | C6H11NO2 |
| Molecular Weight (g/mol) | 129.159 |
| MDL Number | MFCD00019883 |
| InChI Key | XJHNCGVHPLDMRK-UHFFFAOYSA-N |
| Synonym | 4,4-dimethoxybutyronitrile, 3-cyanopropionaldehyde dimethyl acetal, butanenitrile, 4,4-dimethoxy, 3-cyano propionaldehyde dimethyl acetal, 3-cyanopropionaldehydedimethylacetal, cyanopropionaldehyde dimethyl acetal, .beta.-cyanopropionaldehyde dimethylacetal, 4-dimeth-oxybutyronitrile, acmc-1c7xt, ksc494i0f |
| PubChem CID | 84558 |
| IUPAC Name | 4,4-dimethoxybutanenitrile |
| SMILES | COC(CCC#N)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C089125ML
|
TCI America
C089125ML |
25 mL |
Each for $238.60
|
|
|||||
|
C0891250ML
|
TCI America
C0891250ML |
250 mL |
Each for $1,427.87
|
|
|||||
Chemical Identifiers
| 14618-78-1 | |
| 129.159 | |
| XJHNCGVHPLDMRK-UHFFFAOYSA-N | |
| 84558 | |
| COC(CCC#N)OC |
| C6H11NO2 | |
| MFCD00019883 | |
| 4,4-dimethoxybutyronitrile, 3-cyanopropionaldehyde dimethyl acetal, butanenitrile, 4,4-dimethoxy, 3-cyano propionaldehyde dimethyl acetal, 3-cyanopropionaldehydedimethylacetal, cyanopropionaldehyde dimethyl acetal, .beta.-cyanopropionaldehyde dimethylacetal, 4-dimeth-oxybutyronitrile, acmc-1c7xt, ksc494i0f | |
| 4,4-dimethoxybutanenitrile |
Specifications
| 14618-78-1 | |
| 110°C | |
| MFCD00019883 | |
| 4,4-dimethoxybutyronitrile, 3-cyanopropionaldehyde dimethyl acetal, butanenitrile, 4,4-dimethoxy, 3-cyano propionaldehyde dimethyl acetal, 3-cyanopropionaldehydedimethylacetal, cyanopropionaldehyde dimethyl acetal, .beta.-cyanopropionaldehyde dimethylacetal, 4-dimeth-oxybutyronitrile, acmc-1c7xt, ksc494i0f | |
| COC(CCC#N)OC | |
| 129.159 | |
| 129.16 | |
| Liquid |
| Yellow | |
| C6H11NO2 | |
| 25 mL | |
| XJHNCGVHPLDMRK-UHFFFAOYSA-N | |
| 4,4-dimethoxybutanenitrile | |
| 84558 | |
| ≥98.0% (GC) | |
| 3-Cyanopropionaldehyde Dimethyl Acetal |
Safety and Handling
EINECSNumber : (2)-2787
TSCA : Yes