missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanopropionaldehyde Dimethyl Acetal 98.0+%, TCI America™
Supplier: TCI America C0891250ML
Specifications
| 3-Cyanopropionaldehyde Dimethyl Acetal | |
| Yellow | |
| C6H11NO2 | |
| 250 mL | |
| XJHNCGVHPLDMRK-UHFFFAOYSA-N | |
| 4,4-dimethoxybutanenitrile | |
| 84558 | |
| ≥98.0% (GC) |
| 14618-78-1 | |
| 110°C | |
| MFCD00019883 | |
| 4,4-dimethoxybutyronitrile, 3-cyanopropionaldehyde dimethyl acetal, butanenitrile, 4,4-dimethoxy, 3-cyano propionaldehyde dimethyl acetal, 3-cyanopropionaldehydedimethylacetal, cyanopropionaldehyde dimethyl acetal, .beta.-cyanopropionaldehyde dimethylacetal, 4-dimeth-oxybutyronitrile, acmc-1c7xt, ksc494i0f | |
| COC(CCC#N)OC | |
| 129.159 | |
| 129.16 | |
| Liquid |
Chemical Identifiers
| 14618-78-1 | |
| 129.159 | |
| XJHNCGVHPLDMRK-UHFFFAOYSA-N | |
| 84558 | |
| COC(CCC#N)OC |
| C6H11NO2 | |
| MFCD00019883 | |
| 4,4-dimethoxybutyronitrile, 3-cyanopropionaldehyde dimethyl acetal, butanenitrile, 4,4-dimethoxy, 3-cyano propionaldehyde dimethyl acetal, 3-cyanopropionaldehydedimethylacetal, cyanopropionaldehyde dimethyl acetal, .beta.-cyanopropionaldehyde dimethylacetal, 4-dimeth-oxybutyronitrile, acmc-1c7xt, ksc494i0f | |
| 4,4-dimethoxybutanenitrile |
Safety and Handling
EINECSNumber : (2)-2787
TSCA : Yes