CAS RN 14618-78-1
CAS RN 14618-78-1
IUPAC Name:
4,4-dimethoxybutanenitrile
Synonyms:
4,4-dimethoxybutanenitrile
Molecular Weight (g/mol):
129.16
Molecular Formula:
C6H11NO2
InChi Key:
XJHNCGVHPLDMRK-UHFFFAOYSA-N
SMILES:
COC(CCC#N)OC
1
–
1
of
1
results
1
3-Cyanopropionaldehyde Dimethyl Acetal 98.0+%, TCI America™
CAS: 14618-78-1 Molecular Formula: C6H11NO2 Molecular Weight (g/mol): 129.159 MDL Number: MFCD00019883 InChI Key: XJHNCGVHPLDMRK-UHFFFAOYSA-N Synonym: 4,4-dimethoxybutyronitrile,3-cyanopropionaldehyde dimethyl acetal,butanenitrile, 4,4-dimethoxy,3-cyano propionaldehyde dimethyl acetal,3-cyanopropionaldehydedimethylacetal,cyanopropionaldehyde dimethyl acetal,.beta.-cyanopropionaldehyde dimethylacetal,4-dimeth-oxybutyronitrile,acmc-1c7xt,ksc494i0f PubChem CID: 84558 IUPAC Name: 4,4-dimethoxybutanenitrile SMILES: COC(CCC#N)OC