missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanophenol 98.0+%, TCI America™
Supplier: TCI America C158325G
Specifications
| 3-Cyanophenol | |
| 80°C | |
| MFCD00002252 | |
| 3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113 | |
| C1=CC(=CC(=C1)O)C#N | |
| 119.123 | |
| 119.12 | |
| Crystalline Powder |
| 873-62-1 | |
| C7H5NO | |
| 25 g | |
| SGHBRHKBCLLVCI-UHFFFAOYSA-N | |
| 3-hydroxybenzonitrile | |
| 13394 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 873-62-1 | |
| 119.123 | |
| SGHBRHKBCLLVCI-UHFFFAOYSA-N | |
| 13394 | |
| C1=CC(=CC(=C1)O)C#N |
| C7H5NO | |
| MFCD00002252 | |
| 3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113 | |
| 3-hydroxybenzonitrile |
Safety and Handling
TSCA : No