missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenol, 97%

Catalog No. AC110820100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110820100 10 g Glass bottle
AC110820500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110820100 Supplier Thermo Scientific Chemicals Supplier No. 110820100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 873-62-1 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 873-62-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002252
InChI Key SGHBRHKBCLLVCI-UHFFFAOYSA-N
Synonym 3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113
PubChem CID 13394
IUPAC Name 3-hydroxybenzonitrile
SMILES C1=CC(=CC(=C1)O)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 75°C to 80°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NCC6H4OH
Quantity 10 g
Beilstein 10, 141
Formula Weight 119.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanophenol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-845-4
TSCA TSCA

RUO – Research Use Only