missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzyl Chloride 98.0+%, TCI America™

Catalog No. C27515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27515G Supplier TCI America Supplier No. C27515G

Chemical Identifiers

CAS 64407-07-4
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD02684191
InChI Key WRXVOTDGLNPNND-UHFFFAOYSA-N
Synonym 3-(Chloromethyl)benzonitrile, alpha-Chloro-m-tolunitrile
PubChem CID 522126
IUPAC Name 3-(chloromethyl)benzonitrile
SMILES C1=CC(=CC(=C1)CCl)C#N

Specifications

Melting Point 70°C
Color White
Boiling Point 260°C
Quantity 5 g
UN Number 2923
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanobenzyl Chloride