accessibility menu, dialog, popup

UserName

ClN

CAS RN 64407-07-4

IUPAC Name: 3-(chloromethyl)benzonitrile
Synonyms: 3-(chloromethyl)benzonitrile
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: WRXVOTDGLNPNND-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results