missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanobenzoic Acid 98.0+%, TCI America™
$87.60 - $298.25
Chemical Identifiers
| CAS | 1877-72-1 |
|---|---|
| Molecular Formula | C8H5NO2 |
| Molecular Weight (g/mol) | 147.133 |
| MDL Number | MFCD00002486 |
| InChI Key | GYLKKXHEIIFTJH-UHFFFAOYSA-N |
| Synonym | m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid |
| PubChem CID | 15875 |
| IUPAC Name | 3-cyanobenzoic acid |
| SMILES | C1=CC(=CC(=C1)C(=O)O)C#N |
Chemical Identifiers
| 1877-72-1 | |
| 147.133 | |
| GYLKKXHEIIFTJH-UHFFFAOYSA-N | |
| 15875 | |
| C1=CC(=CC(=C1)C(=O)O)C#N |
| C8H5NO2 | |
| MFCD00002486 | |
| m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid | |
| 3-cyanobenzoic acid |
Specifications
| 1877-72-1 | |
| White | |
| MFCD00002486 | |
| m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid | |
| C1=CC(=CC(=C1)C(=O)O)C#N | |
| 147.133 | |
| 147.13 | |
| Crystalline Powder |
| 220°C | |
| C8H5NO2 | |
| 5 g | |
| GYLKKXHEIIFTJH-UHFFFAOYSA-N | |
| 3-cyanobenzoic acid | |
| 15875 | |
| ≥98.0% (GC,T) | |
| 3-Cyanobenzoic Acid |
Safety and Handling
TSCA : No