accessibility menu, dialog, popup

UserName

CAS RN 1877-72-1

IUPAC Name: 3-cyanobenzoic acid
Synonyms: M-cyanobenzoic acid
Molecular Weight (g/mol): 147.13
Molecular Formula: C8H5NO2
InChi Key: GYLKKXHEIIFTJH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)