missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzoic acid, 98%

CAS: 1877-72-1 | C8H5NO2 | 147.13 g/mol

$121.19 - $121.19

Chemical Identifiers

CAS 1877-72-1
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002486
InChI Key GYLKKXHEIIFTJH-UHFFFAOYSA-N
Synonym m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
PubChem CID 15875
IUPAC Name 3-cyanobenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169130100
SDS View Documents
Thermo Scientific Chemicals
169130100
10 g Glass bottle
Each for $121.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1877-72-1
147.13
GYLKKXHEIIFTJH-UHFFFAOYSA-N
15875
C1=CC(=CC(=C1)C(=O)O)C#N
C8H5NO2
MFCD00002486
m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
3-cyanobenzoic acid

Specifications

1877-72-1
100.0
White
97.5% min. (HPLC)
C8H5NO2
MFCD00002486
09, 835
GYLKKXHEIIFTJH-UHFFFAOYSA-N
3-cyanobenzoic acid
15875
98%
3-Cyanobenzoic acid
97.5
217.0°C to 222.0°C
Authentic
Glass bottle
NCC6H4CO2H
10 g
m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
C1=CC(=CC(=C1)C(=O)O)C#N
147.13
147.13
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air

GHS Signal Word: Warning

EINECSNumber : 217-511-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only