missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzoic acid, 98%

Catalog No. AC169130500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169130500 50 g Glass bottle
AC169130100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169130500 Supplier Thermo Scientific Chemicals Supplier No. 169130500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1877-72-1 | C8H5NO2 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1877-72-1
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002486
InChI Key GYLKKXHEIIFTJH-UHFFFAOYSA-N
Synonym m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
PubChem CID 15875
IUPAC Name 3-cyanobenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 217.0°C to 222.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4CO2H
Quantity 50 g
Beilstein 09, 835
Formula Weight 147.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Cyanobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air
EINECSNumber 217-511-1

RUO – Research Use Only