Learn More
3-Chlorothiophenol, 97%
CAS: 2037-31-2 | C6H5ClS | 144.62 g/mol
Supplier: Thermo Scientific Chemicals L0480614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Chlorothiophenol | |
| 1.244 | |
| 90°C (194°F) | |
| 1.6021 | |
| 25 g | |
| 1854087 | |
| 3-chlorothiophenol, m-chlorothiophenol, benzenethiol, 3-chloro, 3-chlorobenzene-1-thiol, m-chlorobenzenethiol, benzenethiol, m-chloro, 3-chloro thiophenol, 3-chloro-benzenethiol, 3-chloromercaptobenzene, m-chlorophenyl mercaptan | |
| SC1=CC=CC(Cl)=C1 | |
| 144.62 | |
| 144.62 |
| 2037-31-2 | |
| 104°C to 105°C (35 mmHg) | |
| C6H5ClS | |
| MFCD00004839 | |
| UN1760 | |
| Air Sensitive | |
| CQJDYPZUDYXHLM-UHFFFAOYSA-N | |
| 3-chlorobenzenethiol | |
| 16257 | |
| 97% |
Chemical Identifiers
| 2037-31-2 | |
| 144.62 | |
| CQJDYPZUDYXHLM-UHFFFAOYSA-N | |
| 16257 | |
| SC1=CC=CC(Cl)=C1 |
| C6H5ClS | |
| MFCD00004839 | |
| 3-chlorothiophenol, m-chlorothiophenol, benzenethiol, 3-chloro, 3-chlorobenzene-1-thiol, m-chlorobenzenethiol, benzenethiol, m-chloro, 3-chloro thiophenol, 3-chloro-benzenethiol, 3-chloromercaptobenzene, m-chlorophenyl mercaptan | |
| 3-chlorobenzenethiol |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H312-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber : 218-010-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only