missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophenol, 97%

Catalog No. p-7044140
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0480603 1 g
AAL0480606 5 g
AAL0480614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0480603 Supplier Thermo Scientific Chemicals Supplier No. L0480603
Only null left

CAS: 2037-31-2 | C6H5ClS | 144.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2037-31-2
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.62
MDL Number MFCD00004839
InChI Key CQJDYPZUDYXHLM-UHFFFAOYSA-N
Synonym 3-chlorothiophenol, m-chlorothiophenol, benzenethiol, 3-chloro, 3-chlorobenzene-1-thiol, m-chlorobenzenethiol, benzenethiol, m-chloro, 3-chloro thiophenol, 3-chloro-benzenethiol, 3-chloromercaptobenzene, m-chlorophenyl mercaptan
PubChem CID 16257
IUPAC Name 3-chlorobenzenethiol
SMILES SC1=CC=CC(Cl)=C1

Specifications

Density 1.244
Boiling Point 104°C to 105°C (35 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.6021
Quantity 1 g
UN Number UN1760
Beilstein 1854087
Sensitivity Air Sensitive
Formula Weight 144.62
Percent Purity 97%
Chemical Name or Material 3-Chlorothiophenol
Show More Show Less
Hazard Category H227-H302-H312-H314-H332-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 218-010-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only