missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenol 98.0+%, TCI America™

Catalog No. p-7124306
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
C024225G 25 g
C0242100G 100 g
C0242500G 500 g
3 options

Catalog No. C024225G

Supplier: TCI America C024225G

Only null left

Chemical Identifiers

CAS 108-43-0
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002256
InChI Key HORNXRXVQWOLPJ-UHFFFAOYSA-N
Synonym m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641
PubChem CID 7933
ChEBI CHEBI:38855
IUPAC Name 3-chlorophenol
SMILES OC1=CC=CC(Cl)=C1

Specifications

Color White-Yellow
Boiling Point 214°C
Quantity 25 g
UN Number 2020
Formula Weight 128.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chlorophenol
EINECSNumber (3)-0895
RTECSNumber SK2450000
TSCA Yes