accessibility menu, dialog, popup

UserName

CAS RN 108-43-0

IUPAC Name: 3-chlorophenol
Synonyms: 3-chlorophenol ccris 641 C6H5ClO
Molecular Weight (g/mol): 128.56
Molecular Formula: C6H5ClO
InChi Key: HORNXRXVQWOLPJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC(Cl)=C1
Boiling Point 
  • (6)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (6)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)