missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenol, 99%, Thermo Scientific Chemicals

Catalog No. AC109811000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109811000 100 g Glass bottle
AC109810050 5 g Glass bottle
AC109810250 25 g Glass bottle
AC109810010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC109811000 Supplier Thermo Scientific Chemicals Supplier No. 109811000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-43-0 | C6H5ClO | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-43-0
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002256
InChI Key HORNXRXVQWOLPJ-UHFFFAOYSA-N
Synonym m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641
PubChem CID 7933
ChEBI CHEBI:38855
IUPAC Name 3-chlorophenol
SMILES OC1=CC=CC(Cl)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33.0°C to 35.0°C
Color Brown to Yellow
Density 1.2180g/mL
Boiling Point 214.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5650 to 1.5670
Linear Formula ClC6H4OH
Quantity 100 g
Beilstein 06,185
Merck Index 15,2157
Specific Gravity 1.218
Solubility Information Solubility in water: 27.7g/l (20°C). Other solubilities: soluble in alcohol,ether,benzene and aq. alkali
Viscosity 11.55 Pas (25°C)
Formula Weight 128.56
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 3-Chlorophenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in
EINECSNumber 203-582-6
RTECSNumber SK2450000
TSCA TSCA

RUO – Research Use Only