missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenol, 98%

Catalog No. p-7021365
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Catalog No. Quantity
AAA1367118 50 g
AAA1367122 100 g
AAA1367130 250 g
3 options

Catalog No. AAA1367118

Supplier: Thermo Scientific Chemicals A1367118

Only null left

CAS: 108-43-0 | C6H5ClO | 128.56 g/mol

3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is an important intermediate in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is also used as a disinfectant. It is an important intermediate in the synthesis of chemicals.

Solubility
Soluble in ethanol and ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 108-43-0
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002256
InChI Key HORNXRXVQWOLPJ-UHFFFAOYSA-N
Synonym m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641
PubChem CID 7933
ChEBI CHEBI:38855
IUPAC Name 3-chlorophenol
SMILES OC1=CC=CC(Cl)=C1

Specifications

Melting Point 30°C to 34°C
Density 1.245
Boiling Point 214°C
Flash Point 120°C (248°F)
Refractive Index 1.566
Quantity 50 g
UN Number UN2020
Beilstein 1634401
Merck Index 14,2154
Solubility Information Soluble in ethanol and ether.
Formula Weight 128.56
Percent Purity ≥98%
Chemical Name or Material 3-Chlorophenol
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber 203-582-6
RTECSNumber SK2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only