Learn More
3-Chlorophenol, 98%
CAS: 108-43-0 | C6H5ClO | 128.56 g/mol
Supplier: Thermo Scientific Chemicals A1367122
Description
3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is an important intermediate in the synthesis of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is also used as a disinfectant. It is an important intermediate in the synthesis of chemicals.
Solubility
Soluble in ethanol and ether.
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 3-Chlorophenol | |
| 30°C to 34°C | |
| 214°C | |
| C6H5ClO | |
| MFCD00002256 | |
| UN2020 | |
| 14,2154 | |
| Soluble in ethanol and ether. | |
| OC1=CC=CC(Cl)=C1 | |
| 128.56 | |
| CHEBI:38855 | |
| ≥98% |
| 108-43-0 | |
| 1.245 | |
| 120°C (248°F) | |
| 1.566 | |
| 100 g | |
| 1634401 | |
| m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 | |
| HORNXRXVQWOLPJ-UHFFFAOYSA-N | |
| 3-chlorophenol | |
| 7933 | |
| 128.56 |
Chemical Identifiers
| 108-43-0 | |
| 128.56 | |
| HORNXRXVQWOLPJ-UHFFFAOYSA-N | |
| 7933 | |
| 3-chlorophenol |
| C6H5ClO | |
| MFCD00002256 | |
| m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 | |
| CHEBI:38855 | |
| OC1=CC=CC(Cl)=C1 |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber : 203-582-6
RTECSNumber : SK2450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only