missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog No. aah6085409
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6085409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6085409 Supplier Thermo Scientific Chemicals Supplier No. H6085409

CAS: 31719-77-4 | Molecular Formula: C8H7ClO2 | Molecular Weight: 170.59 g/mol

3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 31719-77-4
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00191922
InChI Key PBSUMBYSVFTMNG-UHFFFAOYSA-N
Synonym 3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
PubChem CID 241752
IUPAC Name 3-(chloromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC(CCl)=C1

Specifications

Melting Point 136°C to 138°C
Quantity 10 g
UN Number UN3261
Solubility Information Soluble in methanol.
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)benzoic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only