missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzylamine 98.0+%, TCI America™
Supplier: TCI America C108925G
Specifications
| 3-Chlorobenzylamine | |
| Yellow | |
| C7H8ClN | |
| 25 g | |
| 3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a | |
| C1=CC(=CC(=C1)Cl)CN | |
| 141.598 | |
| 141.60 | |
| Liquid |
| 4152-90-3 | |
| 226°C | |
| MFCD00040752 | |
| 2735 | |
| BJFPYGGTDAYECS-UHFFFAOYSA-N | |
| (3-chlorophenyl)methanamine | |
| 77802 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 4152-90-3 | |
| 141.598 | |
| BJFPYGGTDAYECS-UHFFFAOYSA-N | |
| 77802 | |
| C1=CC(=CC(=C1)Cl)CN |
| C7H8ClN | |
| MFCD00040752 | |
| 3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a | |
| (3-chlorophenyl)methanamine |
Safety and Handling
TSCA : Yes