missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzylamine 98.0+%, TCI America™

Catalog No. p-7124787
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10895G 5 g
C108925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10895G Supplier TCI America Supplier No. C10895G

Chemical Identifiers

CAS 4152-90-3
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00040752
InChI Key BJFPYGGTDAYECS-UHFFFAOYSA-N
Synonym 3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a
PubChem CID 77802
IUPAC Name (3-chlorophenyl)methanamine
SMILES C1=CC(=CC(=C1)Cl)CN

Specifications

Color Yellow
Boiling Point 226°C
Quantity 5 g
UN Number 2735
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzylamine