missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzenesulfonamide, 98%

Catalog No. p-7058325
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0834903 1 g
AAL0834906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0834903 Supplier Thermo Scientific Chemicals Supplier No. L0834903
Only null left

CAS: 17260-71-8 | C6H6ClNO2S | 191.63 g/mol

It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 17260-71-8
Molecular Formula C6H6ClNO2S
Molecular Weight (g/mol) 191.63
MDL Number MFCD00051976
InChI Key WSYQJNPRQUFCGL-UHFFFAOYSA-N
Synonym benzenesulfonamide, 3-chloro, 3-chlorobenzene-1-sulfonamide, 3-chlorobenzenesulphonamide, 3-chloro-benzenesulfonamide, m-chlorobenzenesulfonamide, acmc-20a3ig, 3-chlorobenzensulfonamide, 3-chloranylbenzenesulfonamide, benzenesulfonamide,3-chloro, benzenesulfonamide, 3-chloro-9ci
PubChem CID 519377
IUPAC Name 3-chlorobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 144°C to 148°C
Quantity 1 g
Beilstein 2832245
Solubility Information Slightly soluble in water.
Formula Weight 191.64
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzenesulfonamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only