Learn More
3-Chloroanisole, 98+%
CAS: 2845-89-8 | C7H7ClO | 142.582 g/mol
Supplier: Thermo Scientific Chemicals A1732730
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Chloroanisole | |
| 1.168 | |
| 73°C (163°F) | |
| 1.536 | |
| 250 g | |
| 3-chloroanisole, m-chloroanisole, benzene, 1-chloro-3-methoxy, anisole, m-chloro, 3-chloroanisol, meta-chloroanisole, 3-anisolyl chloride, pubchem3615, 3-chloromethoxybenzene, m-methoxyphenyl chloride | |
| COC1=CC(=CC=C1)Cl | |
| 142.582 | |
| 142.59 |
| 2845-89-8 | |
| 192°C to 194°C | |
| C7H7ClO | |
| MFCD00000591 | |
| 2041497 | |
| YUKILTJWFRTXGB-UHFFFAOYSA-N | |
| 1-chloro-3-methoxybenzene | |
| 17833 | |
| ≥98% |
Chemical Identifiers
| 2845-89-8 | |
| 142.582 | |
| YUKILTJWFRTXGB-UHFFFAOYSA-N | |
| 17833 | |
| COC1=CC(=CC=C1)Cl |
| C7H7ClO | |
| MFCD00000591 | |
| 3-chloroanisole, m-chloroanisole, benzene, 1-chloro-3-methoxy, anisole, m-chloro, 3-chloroanisol, meta-chloroanisole, 3-anisolyl chloride, pubchem3615, 3-chloromethoxybenzene, m-methoxyphenyl chloride | |
| 1-chloro-3-methoxybenzene |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 220-642-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only