missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroanisole, 98+%

Catalog No. p-7023601
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1732718 50 g
AAA1732730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1732718 Supplier Thermo Scientific Chemicals Supplier No. A1732718

CAS: 2845-89-8 | C7H7ClO | 142.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2845-89-8
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00000591
InChI Key YUKILTJWFRTXGB-UHFFFAOYSA-N
Synonym 3-chloroanisole, m-chloroanisole, benzene, 1-chloro-3-methoxy, anisole, m-chloro, 3-chloroanisol, meta-chloroanisole, 3-anisolyl chloride, pubchem3615, 3-chloromethoxybenzene, m-methoxyphenyl chloride
PubChem CID 17833
IUPAC Name 1-chloro-3-methoxybenzene
SMILES COC1=CC(=CC=C1)Cl

Specifications

Density 1.168
Boiling Point 192°C to 194°C
Flash Point 73°C (163°F)
Refractive Index 1.536
Quantity 50 g
Beilstein 2041497
Formula Weight 142.59
Percent Purity ≥98%
Chemical Name or Material 3-Chloroanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 220-642-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only