missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-p-anisaldehyde 98.0+%, TCI America™

Catalog No. p-7125176
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17971G 1 g
C17975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17971G Supplier TCI America Supplier No. C17971G
Only null left

Chemical Identifiers

CAS 4903-09-7
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
InChI Key WYVGYYIZXPXHAZ-UHFFFAOYSA-N
Synonym 3-chloroanisaldehyde, 3-chloro-p-anisaldehyde, benzaldehyde, 3-chloro-4-methoxy, 3-chloro-4-methoxybenzenecarbaldehyde, 3-chloro-4-methoxy-benzaldehyde, pubchem2638, acmc-1ap2w, 3-chloro-4-methoxybenzaldehyde
PubChem CID 78619
IUPAC Name 3-chloro-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)Cl

Specifications

Melting Point 58°C
Color White
Quantity 1 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-p-anisaldehyde