missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-fluorobenzyl Bromide 98.0+%, TCI America™
Supplier: TCI America C24745G
Specifications
| 3-Chloro-5-fluorobenzyl Bromide | |
| Yellow | |
| MFCD03788472 | |
| 3265 | |
| PKSAHOPFPPAUOD-UHFFFAOYSA-N | |
| 1-(bromomethyl)-3-chloro-5-fluorobenzene | |
| 2734834 | |
| ≥98.0% (GC) |
| 493024-39-8 | |
| C7H5BrClF | |
| 5 g | |
| 3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene | |
| FC1=CC(Cl)=CC(CBr)=C1 | |
| 223.47 | |
| 223.47 | |
| Liquid |
Chemical Identifiers
| 493024-39-8 | |
| 223.47 | |
| PKSAHOPFPPAUOD-UHFFFAOYSA-N | |
| 2734834 | |
| FC1=CC(Cl)=CC(CBr)=C1 |
| C7H5BrClF | |
| MFCD03788472 | |
| 3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene | |
| 1-(bromomethyl)-3-chloro-5-fluorobenzene |
Safety and Handling
TSCA : No