missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7125481
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24741G 1 g
C24745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24741G Supplier TCI America Supplier No. C24741G

Chemical Identifiers

CAS 493024-39-8
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD03788472
InChI Key PKSAHOPFPPAUOD-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene
PubChem CID 2734834
IUPAC Name 1-(bromomethyl)-3-chloro-5-fluorobenzene
SMILES FC1=CC(Cl)=CC(CBr)=C1

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 223.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-5-fluorobenzyl Bromide