missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
Supplier: TCI America C33671G
Specifications
| 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 82°C | |
| C11H15BClNO2 | |
| 1 g | |
| WCKBNAJTZUVHPD-UHFFFAOYSA-N | |
| 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 23157866 | |
| ≥98.0% (GC,T) |
| 865186-94-3 | |
| White-Yellow | |
| MFCD06798284 | |
| 5-chloropyridine-3-boronic acid pinacol ester, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloropyridine-5-boronic acid pinacol ester, 3-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 5-chloropyridine-3-boronic acid, pinacol ester, 5-chloropyridin-3-ylboronic acid pinacol ester, pubchem21018, abbypharma ap-12-5241 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)Cl | |
| 239.506 | |
| 239.51 | |
| Crystalline Powder |
Chemical Identifiers
| 865186-94-3 | |
| 239.506 | |
| WCKBNAJTZUVHPD-UHFFFAOYSA-N | |
| 23157866 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)Cl |
| C11H15BClNO2 | |
| MFCD06798284 | |
| 5-chloropyridine-3-boronic acid pinacol ester, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloropyridine-5-boronic acid pinacol ester, 3-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 5-chloropyridine-3-boronic acid, pinacol ester, 5-chloropyridin-3-ylboronic acid pinacol ester, pubchem21018, abbypharma ap-12-5241 | |
| 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
TSCA : No