accessibility menu, dialog, popup

UserName

CAS RN 865186-94-3

BOOH3CCH3H3CH3CNCl

CAS RN 865186-94-3

IUPAC Name: 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 239.51
Molecular Formula: C11H15BClNO2
InChi Key: WCKBNAJTZUVHPD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(Cl)=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results