missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. p-7125941
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3367200MG 200 mg
C33671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3367200MG Supplier TCI America Supplier No. C3367200MG
Only null left

Chemical Identifiers

CAS 865186-94-3
Molecular Formula C11H15BClNO2
Molecular Weight (g/mol) 239.506
MDL Number MFCD06798284
InChI Key WCKBNAJTZUVHPD-UHFFFAOYSA-N
Synonym 5-chloropyridine-3-boronic acid pinacol ester, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloropyridine-5-boronic acid pinacol ester, 3-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 5-chloropyridine-3-boronic acid, pinacol ester, 5-chloropyridin-3-ylboronic acid pinacol ester, pubchem21018, abbypharma ap-12-5241
PubChem CID 23157866
IUPAC Name 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)Cl

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 200 mg
Formula Weight 239.51
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine