missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethyl)benzoic acid, 97%

Catalog No. p-7031091
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAH3203803 1 g
AAH3203806 5 g
2 options

Catalog No. AAH3203803

Supplier: Thermo Scientific Chemicals H3203803

Only null left

CAS: 115754-20-6 | C8H4ClF3O2 | 224.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115754-20-6
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD04972755
InChI Key UDXPRKSPAZWHQN-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethyl benzoic acid, benzoic acid, 3-chloro-4-trifluoromethyl, 3-chloro-4-trifluoromethyl-benzoic acid, acmc-1c5tn, chloro-4-trifluoromethyl benzoic acid, 3-chloro-4-trifluoromethyl benzoic acid, jrd
PubChem CID 21180297
IUPAC Name 3-chloro-4-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(Cl)=C(C=C1)C(F)(F)F

Specifications

Melting Point 158°C to 161°C
Quantity 1 g
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only