CAS RN 115754-20-6
IUPAC Name:
3-chloro-4-(trifluoromethyl)benzoic acid
Synonyms:
3-chloro-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
224.56
Molecular Formula:
C8H4ClF3O2
InChi Key:
UDXPRKSPAZWHQN-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(Cl)=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
3-Chloro-4-(trifluoromethyl)benzoic acid, 97%
3-Chloro-4-(trifluoromethyl)benzoic acid, 97%
CAS: 115754-20-6 Molecular Formula: C8H4ClF3O2 Molecular Weight (g/mol): 224.56 MDL Number: MFCD04972755 InChI Key: UDXPRKSPAZWHQN-UHFFFAOYSA-N Synonym: 3-chloro-4-trifluoromethyl benzoic acid,benzoic acid, 3-chloro-4-trifluoromethyl,3-chloro-4-trifluoromethyl-benzoic acid,acmc-1c5tn,chloro-4-trifluoromethyl benzoic acid,3-chloro-4-trifluoromethyl benzoic acid, jrd PubChem CID: 21180297 IUPAC Name: 3-chloro-4-(trifluoromethyl)benzoic acid SMILES: OC(=O)C1=CC(Cl)=C(C=C1)C(F)(F)F