missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethyl)benzoic acid, 97%

Catalog No. AAH3203803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3203803 1 g
AAH3203806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3203803 Supplier Thermo Scientific Chemicals Supplier No. H3203803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 115754-20-6 | C8H4ClF3O2 | 224.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115754-20-6
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD04972755
InChI Key UDXPRKSPAZWHQN-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethyl benzoic acid, benzoic acid, 3-chloro-4-trifluoromethyl, 3-chloro-4-trifluoromethyl-benzoic acid, acmc-1c5tn, chloro-4-trifluoromethyl benzoic acid, 3-chloro-4-trifluoromethyl benzoic acid, jrd
PubChem CID 21180297
IUPAC Name 3-chloro-4-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(Cl)=C(C=C1)C(F)(F)F

Specifications

Melting Point 158°C to 161°C
Quantity 1 g
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only