missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-nitrotoluene 98.0+%, TCI America™
$48.46 - $141.67
Chemical Identifiers
| CAS | 38939-88-7 |
|---|---|
| Molecular Formula | C7H6ClNO2 |
| Molecular Weight (g/mol) | 171.58 |
| MDL Number | MFCD02683043 |
| InChI Key | KGSQRFPDZCBVBS-UHFFFAOYSA-N |
| Synonym | 3-chloro-4-nitrotoluene, 2-chloro-4-methyl-1-nitro-benzene, benzene, 2-chloro-4-methyl-1-nitro, pubchem4534, acmc-1afh8, ksc493s6j |
| PubChem CID | 123478 |
| IUPAC Name | 2-chloro-4-methyl-1-nitrobenzene |
| SMILES | CC1=CC(=C(C=C1)[N+](=O)[O-])Cl |
Chemical Identifiers
| 38939-88-7 | |
| 171.58 | |
| KGSQRFPDZCBVBS-UHFFFAOYSA-N | |
| 123478 | |
| CC1=CC(=C(C=C1)[N+](=O)[O-])Cl |
| C7H6ClNO2 | |
| MFCD02683043 | |
| 3-chloro-4-nitrotoluene, 2-chloro-4-methyl-1-nitro-benzene, benzene, 2-chloro-4-methyl-1-nitro, pubchem4534, acmc-1afh8, ksc493s6j | |
| 2-chloro-4-methyl-1-nitrobenzene |
Specifications
| 38939-88-7 | |
| 101°C | |
| MFCD02683043 | |
| 3457 | |
| KGSQRFPDZCBVBS-UHFFFAOYSA-N | |
| 2-chloro-4-methyl-1-nitrobenzene | |
| 123478 | |
| ≥98.0% (GC) | |
| 3-Chloro-4-nitrotoluene |
| White-Yellow | |
| C7H6ClNO2 | |
| 5 g | |
| 3-chloro-4-nitrotoluene, 2-chloro-4-methyl-1-nitro-benzene, benzene, 2-chloro-4-methyl-1-nitro, pubchem4534, acmc-1afh8, ksc493s6j | |
| CC1=CC(=C(C=C1)[N+](=O)[O-])Cl | |
| 171.58 | |
| 171.58 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (3)-0449
TSCA : No