CAS RN 38939-88-7
CAS RN 38939-88-7
IUPAC Name:
2-chloro-4-methyl-1-nitrobenzene
Synonyms:
3-chloro-4-nitrotoluene
Molecular Weight (g/mol):
171.58
Molecular Formula:
C7H6ClNO2
InChi Key:
KGSQRFPDZCBVBS-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C(Cl)=C1)[N+]([O-])=O
1
–
1
of
1
results
1
3-Chloro-4-nitrotoluene 98.0+%, TCI America™
CAS: 38939-88-7 Molecular Formula: C7H6ClNO2 Molecular Weight (g/mol): 171.58 MDL Number: MFCD02683043 InChI Key: KGSQRFPDZCBVBS-UHFFFAOYSA-N Synonym: 3-chloro-4-nitrotoluene,2-chloro-4-methyl-1-nitro-benzene,benzene, 2-chloro-4-methyl-1-nitro,pubchem4534,acmc-1afh8,ksc493s6j PubChem CID: 123478 IUPAC Name: 2-chloro-4-methyl-1-nitrobenzene SMILES: CC1=CC(=C(C=C1)[N+](=O)[O-])Cl