missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-nitrotoluene 98.0+%, TCI America™

Catalog No. C22775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22775G 5 g
C227725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22775G Supplier TCI America Supplier No. C22775G
Only null left

Chemical Identifiers

CAS 38939-88-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD02683043
InChI Key KGSQRFPDZCBVBS-UHFFFAOYSA-N
Synonym 3-chloro-4-nitrotoluene, 2-chloro-4-methyl-1-nitro-benzene, benzene, 2-chloro-4-methyl-1-nitro, pubchem4534, acmc-1afh8, ksc493s6j
PubChem CID 123478
IUPAC Name 2-chloro-4-methyl-1-nitrobenzene
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])Cl

Specifications

Color White-Yellow
Boiling Point 101°C
Quantity 5 g
UN Number 3457
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chloro-4-nitrotoluene
EINECSNumber (3)-0449
TSCA No