missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylaniline 97.0+%, TCI America™

Catalog No. C053825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C053825G 25 g
C0538500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C053825G Supplier TCI America Supplier No. C053825G

Chemical Identifiers

CAS 95-74-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007773
InChI Key RQKFYFNZSHWXAW-UHFFFAOYSA-N
Synonym 3-chloro-p-toluidine, 2-chloro-4-aminotoluene, 4-amino-2-chlorotoluene, benzenamine, 3-chloro-4-methyl, 3-chloro-4-methylbenzenamine, 3-chloro-4-methylphenylamine, 1-amino-3-chloro-4-methylbenzene, 4-methyl-3-chloroaniline, p-toluidine, 3-chloro, gull toxicant
PubChem CID 7255
ChEBI CHEBI:37824
IUPAC Name 3-chloro-4-methylaniline
SMILES CC1=C(C=C(C=C1)N)Cl

Specifications

Color White-Yellow
Boiling Point 244°C
Quantity 25 g
UN Number 2239
Formula Weight 141.60
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chloro-4-methylaniline
EINECSNumber (3)-0265
RTECSNumber XU5111000
TSCA Yes