missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methoxybenzoic Acid 97.0+%, TCI America™

Catalog No. p-7125523
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25505G Supplier TCI America Supplier No. C25505G

Chemical Identifiers

CAS 37908-96-6
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00016512
InChI Key IBCQUQXCTOPJOD-UHFFFAOYSA-N
Synonym 3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid
PubChem CID 169982
IUPAC Name 3-chloro-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)Cl

Specifications

Melting Point 217°C
Quantity 5 g
Formula Weight 186.59
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-methoxybenzoic Acid