missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-iodobenzotrifluoride (stabilized with Copper chip) 97.0+%, TCI America™
$46.60 - $136.08
Chemical Identifiers
| CAS | 141738-80-9 |
|---|---|
| Molecular Formula | C7H3ClF3I |
| Molecular Weight (g/mol) | 306.45 |
| MDL Number | MFCD00042562 |
| InChI Key | ULJRBVGSKAKZKQ-UHFFFAOYSA-N |
| Synonym | 3-chloro-4-iodobenzotrifluoride, 2-chloro-1-iodo-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl iodobenzene, benzene, 2-chloro-1-iodo-4-trifluoromethyl, pubchem1973, acmc-209cms, buttpark 9857-21, 2-chloro-4-trifluoromethyliodobenzene |
| PubChem CID | 2736612 |
| IUPAC Name | 2-chloro-1-iodo-4-(trifluoromethyl)benzene |
| SMILES | FC(F)(F)C1=CC=C(I)C(Cl)=C1 |
Chemical Identifiers
| 141738-80-9 | |
| 306.45 | |
| ULJRBVGSKAKZKQ-UHFFFAOYSA-N | |
| 2736612 | |
| FC(F)(F)C1=CC=C(I)C(Cl)=C1 |
| C7H3ClF3I | |
| MFCD00042562 | |
| 3-chloro-4-iodobenzotrifluoride, 2-chloro-1-iodo-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl iodobenzene, benzene, 2-chloro-1-iodo-4-trifluoromethyl, pubchem1973, acmc-209cms, buttpark 9857-21, 2-chloro-4-trifluoromethyliodobenzene | |
| 2-chloro-1-iodo-4-(trifluoromethyl)benzene |
Specifications
| 141738-80-9 | |
| 60°C | |
| MFCD00042562 | |
| 3-chloro-4-iodobenzotrifluoride, 2-chloro-1-iodo-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl iodobenzene, benzene, 2-chloro-1-iodo-4-trifluoromethyl, pubchem1973, acmc-209cms, buttpark 9857-21, 2-chloro-4-trifluoromethyliodobenzene | |
| FC(F)(F)C1=CC=C(I)C(Cl)=C1 | |
| 306.45 | |
| 306.45 | |
| Liquid |
| Yellow | |
| C7H3ClF3I | |
| 1 g | |
| ULJRBVGSKAKZKQ-UHFFFAOYSA-N | |
| 2-chloro-1-iodo-4-(trifluoromethyl)benzene | |
| 2736612 | |
| ≥97.0% (GC) | |
| 3-Chloro-4-iodobenzotrifluoride (stabilized with Copper chip) |
Safety and Handling
TSCA : No