CAS RN 141738-80-9
IUPAC Name:
2-chloro-1-iodo-4-(trifluoromethyl)benzene
Synonyms:
2-chloro-1-iodo-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
306.45
Molecular Formula:
C7H3ClF3I
InChi Key:
ULJRBVGSKAKZKQ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(I)C(Cl)=C1
1
–
1
of
1
results
1
3-Chloro-4-iodobenzotrifluoride (stabilized with Copper chip) 97.0+%, TCI America™
CAS: 141738-80-9 Molecular Formula: C7H3ClF3I Molecular Weight (g/mol): 306.45 MDL Number: MFCD00042562 InChI Key: ULJRBVGSKAKZKQ-UHFFFAOYSA-N Synonym: 3-chloro-4-iodobenzotrifluoride,2-chloro-1-iodo-4-trifluoromethyl benzene,2-chloro-4-trifluoromethyl iodobenzene,benzene, 2-chloro-1-iodo-4-trifluoromethyl,pubchem1973,acmc-209cms,buttpark 9857-21,2-chloro-4-trifluoromethyliodobenzene PubChem CID: 2736612 IUPAC Name: 2-chloro-1-iodo-4-(trifluoromethyl)benzene SMILES: FC(F)(F)C1=CC=C(I)C(Cl)=C1