missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-iodobenzotrifluoride (stabilized with Copper chip) 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 60°C |
| Quantity | 5 g |
| Formula Weight | 306.45 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Chloro-4-iodobenzotrifluoride (stabilized with Copper chip) |
Safety and Handling
| TSCA | No |