missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-iodoaniline 95.0+%, TCI America™

Catalog No. C21675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21675G 5 g
C216725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21675G Supplier TCI America Supplier No. C21675G

Chemical Identifiers

CAS 135050-44-1
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.47
MDL Number MFCD00051845
InChI Key ONZHMGRKWJMTDE-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-iodo, 3-chloro-4-iodobenzenamine, 4-amino-2-chloroiodobenzene, 3-chloro-4-iodophenylamine, pubchem3812, 3-chloro4-iodoaniline, zlchem 1106, acmc-209bxp, 3-chloro-4-iodo-aniline, 3-chloro-4-iodo aniline
PubChem CID 282926
IUPAC Name 3-chloro-4-iodoaniline
SMILES NC1=CC=C(I)C(Cl)=C1

Specifications

Melting Point 70°C
Color Purple-White
Quantity 5 g
Formula Weight 253.47
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-iodoaniline